1,3-Benzodioxole-4-carbaldehyde ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₆O₃ |
MDL Number:
MFCD00016902 CAS Number: 7797-83-3 EINECS: 232-252-4 |
|
Risultati del test di specificazione
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...