Stampa…

4-Methylbenzyl phenyl ketone ≥97.0% (by GC)

Fornitore: TCI

M1639-25G M1639-5G
TCIAM1639-25GEA 151 EUR
TCIAM1639-25G TCIAM1639-5G
4-Methylbenzyl phenyl ketone ≥97.0% (by GC)
4-Methylbenzyl phenyl ketone
Formula: C₁₅H₁₄O
Punto di ebollizione: 155°C
MDL Number: MFCD00017513
CAS Number: 2430-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd458710204b95c639c2386c2ba4b517d8d3e91c&vid=cd458710204b95c639c2386c2ba4b517d8d3e91c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR