Stampa…

(Triphenylphosphoranylidene)ketene

Fornitore: TCI EUROPE

Synonyms: Bestmann Ylide, Ketenylidene(triphenyl)phosphorane

T2565-1G T2565-5G
TCIAT2565-1GEA 72 EUR
TCIAT2565-1G TCIAT2565-5G
(Triphenylphosphoranylidene)ketene
(Triphenylphosphoranylidene)ketene
Formula: C₂₀H₁₅OP
MDL Number: MFCD00040613
CAS Number: 15596-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9effb0ed4c1277dfdad2162aede07c7041896f4&vid=e9effb0ed4c1277dfdad2162aede07c7041896f4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Melting point 168.0 to 172.0 deg-C
Elemental analysis (Carbon) 75.50 to 83.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR