Stampa…

Dihydroergotamine mesylate ≥97.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dihydroergotamine methanesulphonate

D2633-100MG D2633-1G
TCIAD2633-100MGEA 30 EUR
TCIAD2633-100MG TCIAD2633-1G
Dihydroergotamine mesylate ≥97.0% (by HPLC)
Dihydroergotamine mesylate
Formula: C₃₄H₄₁N₅O₈S
Peso molecolare: 679,78 g/mol
MDL Number: MFCD00058615
CAS Number: 6190-39-2
EINECS: 228-235-6
Merck Index: 14,03174
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6013e050868dac39f100a7c074fa400c8950364d&vid=6013e050868dac39f100a7c074fa400c8950364d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D -17.0 to -23.0 deg (C=0.25, JP Method) (calcd.on dried substance)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR