Stampa…

α,α-Diphenyl-N-methyl-D-prolinol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

H0784-1G H0784-5G H0784-100MG
TCIAH0784-1GEA 167 EUR
TCIAH0784-1G TCIAH0784-5G TCIAH0784-100MG
α,α-Diphenyl-N-methyl-D-prolinol ≥98.0% (by GC)
α,α-Diphenyl-N-methyl-D-prolinol
Formula: C₁₈H₂₁NO
MDL Number: MFCD00145217
CAS Number: 144119-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99a5295f672ffa691f8dda7ff597852788be0257&vid=99a5295f672ffa691f8dda7ff597852788be0257&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 73.0 deg-C
Specific rotation [a]20/D -53.0 to -58.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR