Stampa…

(S)-(-)-α,α-Diphenylprolinol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol

D2735-1G D2735-5G
TCIAD2735-1GEA 41 EUR
TCIAD2735-1G TCIAD2735-5G
(S)-(-)-α,α-Diphenylprolinol ≥98.0% (by GC, titration analysis)
(S)-(-)-α,α-Diphenylprolinol
Formula: C₁₇H₁₉NO
Peso molecolare: 253,35 g/mol
MDL Number: MFCD00075506
CAS Number: 112068-01-6
Merck Index: 14,00155
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d009940ccbf6f711fa7869077f1882d797768e&vid=f2d009940ccbf6f711fa7869077f1882d797768e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 99.5 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR