Triphenylmethyl mercaptan ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Triphenylmethanethiol
T2244-25G
T2244-5G
TCIAT2244-25GEA
118
EUR
TCIAT2244-25G
TCIAT2244-5G
Triphenylmethyl mercaptan ≥97.0% (by HPLC)
Triphenylmethyl mercaptan
Formula:
C₁₉H₁₆S Peso molecolare: 276,40 g/mol |
MDL Number:
MFCD00004854 CAS Number: 3695-77-0 EINECS: 223-020-3 |
Risultati del test di specificazione
Purity (HPLC) | min. 97.0 area% |
Melting point | 106.0 to 110.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...