Stampa…

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate ≥95.0% (by GC)

Fornitore: TCI
Pericolo

I0398-1G I0398-5G
TCIAI0398-1GEA 70 EUR
TCIAI0398-1G TCIAI0398-5G
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate ≥95.0% (by GC)
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate
Formula: C₁₃H₁₁NO
Flash Pt: 93°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00064181
CAS Number: 73671-79-1
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ef4435d2cc61463e80d8745453dbdf689d3aff2&vid=7ef4435d2cc61463e80d8745453dbdf689d3aff2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid to slightly cloudy liquid
Purity (GC) min. 95.0 %
Optical purity (GC) min. 97.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR