(R)-(-)-1-(1-Naphthyl)ethyl isocyanate ≥90.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
I0336-1G
I0336-5G
TCIAI0336-1GEA
72
EUR
TCIAI0336-1G
TCIAI0336-5G
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate ≥90.0% (by GC)
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate
Formula:
C₁₃H₁₁NO Peso molecolare: 197,24 g/mol Flash Pt: 93°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00004015 CAS Number: 42340-98-7 EINECS: 255-759-2 UN: 2206 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 90.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | -40.0 to -50.0 deg (C=5, Toluene) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...