Stampa…

Mucobromic acid ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2,3-Dibromomalealdehydic acid, (Z)-2,3-Dibromo-4-oxobut-2-enoic acid

M0471-25G M0471-500G
TCIAM0471-25GEA 62 EUR
TCIAM0471-25G TCIAM0471-500G
Mucobromic acid ≥99.0% (by titrimetric analysis)
Mucobromic acid
Formula: C₄H₂Br₂O₃
Peso molecolare: 257,88 g/mol
MDL Number: MFCD00063745
CAS Number: 488-11-9
EINECS: 207-670-5
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1913b25c64325a6577ef51945889fdc167bc80ea&vid=1913b25c64325a6577ef51945889fdc167bc80ea&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 123.0 to 126.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR