Stampa…

Acetohydroxamic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: N-Hydroxyacetamide

A0051-10G A0051-25G
TCIAA0051-10GEA 72 EUR
TCIAA0051-10G TCIAA0051-25G
Acetohydroxamic acid ≥98.0% (by titrimetric analysis)
Acetohydroxamic acid
Formula: C₂H₅NO₂
Peso molecolare: 75.0666 g/mol
MDL Number: MFCD00009994
CAS Number: 546-88-3
EINECS: 208-913-8
Merck Index: 14,00063
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eeecd0eaecef85da56b7040c3bd71486c620f290&vid=eeecd0eaecef85da56b7040c3bd71486c620f290&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 87.0 to 92.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR