Stampa…

tert-Butyl ethyl malonate ≥95.0% (by GC)

Fornitore: TCI EUROPE

B3720-25G B3720-5G
TCIAB3720-25GEA 97 EUR
TCIAB3720-25G TCIAB3720-5G
tert-Butyl ethyl malonate ≥95.0% (by GC)
tert-Butyl ethyl malonate
Formula: C₉H₁₆O₄
Peso molecolare: 188,22 g/mol
Punto di ebollizione: 84°C
Flash Pt: 87°C
MDL Number: MFCD00009193
CAS Number: 32864-38-3
EINECS: 251-269-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71295f8d2674f6275a7e81862b717b3766d424ae&vid=71295f8d2674f6275a7e81862b717b3766d424ae&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR