2-Amino-3-chloropyrazine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
A2143-1G
A2143-5G
TCIAA2143-1GEA
27
EUR
TCIAA2143-1G
TCIAA2143-5G
2-Amino-3-chloropyrazine ≥98.0% (by GC)
2-Amino-3-chloropyrazine
Formula:
C₄H₄ClN₃ |
MDL Number:
MFCD04114305 CAS Number: 6863-73-6 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...