Stampa…

Benzyltrimethylammonium dichloroiodate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

B1604-25G B1604-5G
TCIAB1604-25GEA 63 EUR
TCIAB1604-25G TCIAB1604-5G
Benzyltrimethylammonium dichloroiodate ≥97.0% (by titrimetric analysis)
Benzyltrimethylammonium dichloroiodate
Formula: C₁₀H₁₆Cl₂IN
Peso molecolare: 348,06 g/mol
MDL Number: MFCD00075259
CAS Number: 114971-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc7b9bbf3d6f19ab4363e2ad4ffe438d35b4f679&vid=dc7b9bbf3d6f19ab4363e2ad4ffe438d35b4f679&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 125.0 to 128.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR