Stampa…

Diethyl mesoxalate (diethyl ketomalonate) ≥94.0% (by GC)

Fornitore: TCI

M0866-5G M0866-25G
TCIAM0866-5GEA 58 EUR
TCIAM0866-5G TCIAM0866-25G
Diethyl mesoxalate (diethyl ketomalonate) ≥94.0% (by GC)
Diethyl mesoxalate (diethyl ketomalonate)
Formula: C₇H₁₀O₅
Punto di ebollizione: 210°C
MDL Number: MFCD00009121
CAS Number: 609-09-6
EINECS: 210-176-2
Merck Index: 14,05913
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f0814e527d647112871cc3e01f84fcb7764c00&vid=b8f0814e527d647112871cc3e01f84fcb7764c00&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR