3-(Methylthio)propionic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-(Methylthio)propionic Acid
M0811-25G
M0811-100G
TCIAM0811-25GEA
36
EUR
TCIAM0811-25G
TCIAM0811-100G
3-(Methylthio)propionic acid ≥98.0% (by GC, titration analysis)
3-(Methylthio)propionic acid
Formula:
C₄H₈O₂S Peso molecolare: 120,17 g/mol Punto di ebollizione: 130°C Flash Pt: 104°C |
MDL Number:
MFCD00059635 CAS Number: 646-01-5 EINECS: 211-460-9 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...