Methyl-α-D-glucopyranoside ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
M0228-25G
M0228-500G
M0228-100G
TCIAM0228-25GEA
17
EUR
TCIAM0228-25G
TCIAM0228-500G
TCIAM0228-100G
Methyl-α-D-glucopyranoside ≥98.0% (by GC)
Methyl-α-D-glucopyranoside
Formula:
C₇H₁₄O₆ Peso molecolare: 194,18 g/mol |
MDL Number:
MFCD00064086 CAS Number: 97-30-3 EINECS: 202-571-3 Merck Index: 14,06080 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 167.0 to 170.0 deg-C |
Specific rotation [a]20/D | +157.0 to +162.0 deg (C=10, H2O) |
Solubility in Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...