Ethyl cyclohexylacetate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Cyclohexylacetic acid ethyl ester, Ethyl Cyclohexaneacetate, Cyclohexaneacetic acid ethyl ester
C1076-25ML
C1076-500ML
TCIAC1076-25MLEA
52
EUR
TCIAC1076-25ML
TCIAC1076-500ML
Ethyl cyclohexylacetate ≥98.0% (by GC)
Ethyl cyclohexylacetate
Formula:
C₁₀H₁₈O₂ Punto di ebollizione: 212°C Flash Pt: 80°C |
MDL Number:
MFCD00001517 CAS Number: 5452-75-5 EINECS: 226-695-2 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...