2-Isopropyl-3-methoxypyrazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
I0577-1G
I0577-5G
TCIAI0577-1GEA
47
EUR
TCIAI0577-1G
TCIAI0577-5G
2-Isopropyl-3-methoxypyrazine ≥98.0% (by GC)
2-Isopropyl-3-methoxypyrazine
Formula:
C₈H₁₂N₂O Peso molecolare: 152,1900 g/mol Flash Pt: 67°C |
MDL Number:
MFCD00006135 CAS Number: 25773-40-4 EINECS: 247-256-1 |
Risultati del test di specificazione
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...