Stampa…

Ethyl phenylglyoxylate ≥96.0% (by GC)

Fornitore: TCI

B0109-25ML B0109-5ML
TCIAB0109-25MLEA 66 EUR
TCIAB0109-25ML TCIAB0109-5ML
Ethyl phenylglyoxylate ≥96.0% (by GC)
Ethyl phenylglyoxylate
Formula: C₁₀H₁₀O₃
MDL Number: MFCD00009120
CAS Number: 1603-79-8
EINECS: 216-504-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea03bfacc6c4fd8c2fde2e99e3d9769e6edcaaf7&vid=ea03bfacc6c4fd8c2fde2e99e3d9769e6edcaaf7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR