Stampa…

Tripalmitin (glycerol tripalmitate) ≥85.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate

G0213-25G G0213-250G
TCIAG0213-25GEA 82 EUR
TCIAG0213-25G TCIAG0213-250G
Tripalmitin (glycerol tripalmitate) ≥85.0% (by GC)
Tripalmitin (glycerol tripalmitate)
Formula: C₅₁H₉₈O₆
Peso molecolare: 807,34 g/mol
Punto di ebollizione: 320°C
MDL Number: MFCD00008995
CAS Number: 555-44-2
EINECS: 209-098-1
Merck Index: 14,09736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e4b82c4e429e24451fdef8e13873e8c26ad61e3&vid=2e4b82c4e429e24451fdef8e13873e8c26ad61e3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 85.0 %
Melting point 64.0 to 68.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR