Stampa…

1(2H)-Phthalazinone ≥98.0% (by GC)

Fornitore: TCI
Attenzione

P1179-25G P1179-500G
TCIAP1179-25GEA 24 EUR
TCIAP1179-25G TCIAP1179-500G
1(2H)-Phthalazinone ≥98.0% (by GC)
1(2H)-Phthalazinone
Formula: C₈H₆N₂O
Punto di ebollizione: 337°C
MDL Number: MFCD00006892
CAS Number: 119-39-1
EINECS: 204-319-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63f08aca08fe9336ffce8687223ed8fd6cc05c88&vid=63f08aca08fe9336ffce8687223ed8fd6cc05c88&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 183.0 to 187.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR