Stampa…

3-(2-Benzothiazolyl)-7-(diethylamino)coumarin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Benzo[d]thiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one, Coumarin 6

B2088-1G B2088-5G
TCIAB2088-1GEA 52 EUR
TCIAB2088-1G TCIAB2088-5G
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin ≥98.0% (by HPLC, titration analysis)
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin
Formula: C₂₀H₁₈N₂O₂S
Peso molecolare: 350,4341 g/mol
MDL Number: MFCD00041869
CAS Number: 38215-36-0
EINECS: 253-830-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0b3221a5c293a551d900bf62c121ca0ec35ffe1&vid=b0b3221a5c293a551d900bf62c121ca0ec35ffe1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 206.0 to 210.0 deg-C
Molar extinction coefficient min. 52000 (THF, 441.0 to 445.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR