Stampa…

L(-)-Carnitine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

C0049-25G C0049-5G
TCIAC0049-25GEA 123 EUR
TCIAC0049-25G TCIAC0049-5G
L(-)-Carnitine ≥98.0% (by HPLC, titration analysis)
L(-)-Carnitine
Formula: C₇H₁₅NO₃
Peso molecolare: 161,20 g/mol
MDL Number: MFCD00038747
CAS Number: 541-15-1
EINECS: 208-768-0
Merck Index: 14,01849
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fdc3ce56dcb7b3b5534172567328653c3d1b793&vid=3fdc3ce56dcb7b3b5534172567328653c3d1b793&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -30.0 to -33.0 deg (C=1, H2O)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR