(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(-)-2,3-O-Isopropylidene-D-threitol
I0375-1G
I0375-5G
TCIAI0375-1GEA
78
EUR
TCIAI0375-1G
TCIAI0375-5G
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol ≥98.0% (by GC)
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol
Formula:
C₇H₁₄O₄ Peso molecolare: 162,18 g/mol Punto di ebollizione: 96°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00009761 CAS Number: 73346-74-4 EINECS: 277-391-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 52.0 deg-C |
Specific rotation [a]20/D | -3.0 to -5.0 deg (C=5, Aceton) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...