Stampa…

Diethyl phosphoramidate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Diethyl phosphoryl Amide, Diethylphosphoramidate, Diethyl Amidophosphate

D2361-25G D2361-5G
TCIAD2361-25GEA 109 EUR
TCIAD2361-25G TCIAD2361-5G
Diethyl phosphoramidate ≥98.0% (by GC)
Diethyl phosphoramidate
Formula: C₄H₁₂NO₃P
Peso molecolare: 153,1167 g/mol
Punto di ebollizione: 140°C
MDL Number: MFCD00015676
CAS Number: 1068-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f4be8115dbf39dd6e1694c9a9786882bab62262&vid=1f4be8115dbf39dd6e1694c9a9786882bab62262&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR