8-Quinolinesulphonyl chloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₉H₆ClNO₂S |
MDL Number:
MFCD00006808 CAS Number: 18704-37-5 EINECS: 242-515-5 UN: 3261 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...