Stampa…

3-Chlorobenzo[b]thiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C2444-1G C2444-5G
TCIAC2444-1GEA 103 EUR
TCIAC2444-1G TCIAC2444-5G
3-Chlorobenzo[b]thiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
3-Chlorobenzo[b]thiophene-2-carboxylic acid
Formula: C₉H₅ClO₂S
Peso molecolare: 212,65 g/mol
Punto di fusione: 271°C
MDL Number: MFCD00014579
CAS Number: 21211-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b0eead7fe3b91607136151a8b23c3c2d856e51d&vid=2b0eead7fe3b91607136151a8b23c3c2d856e51d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR