Stampa…

N,N-Diethylhydroxylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D1339-25ML D1339-500ML D1339-100ML
TCIAD1339-25MLEA 18 EUR
TCIAD1339-25ML TCIAD1339-500ML TCIAD1339-100ML
N,N-Diethylhydroxylamine ≥98.0% (by GC, titration analysis)
N,N-Diethylhydroxylamine
Formula: C₄H₁₁NO
Peso molecolare: 89,14 g/mol
Punto di ebollizione: 122°C
Flash Pt: 51°C
MDL Number: MFCD00002126
CAS Number: 3710-84-7
EINECS: 223-055-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5983be6139da5517027b116de8c1253f058dc6b9&vid=5983be6139da5517027b116de8c1253f058dc6b9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR