Iminodiacetic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,2'-Azanediyldiacetic acid
I0006-25G
I0006-500G
TCIAI0006-25GEA
16
EUR
TCIAI0006-25G
TCIAI0006-500G
Iminodiacetic acid ≥98.0% (by titrimetric analysis)
Iminodiacetic acid
Formula:
C₄H₇NO₄ Peso molecolare: 133,10 g/mol |
MDL Number:
MFCD00004280 CAS Number: 142-73-4 EINECS: 205-555-4 Merck Index: 14,04917 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...