Stampa…

Daminozide (succinic acid mono(2,2-dimethylhydrazide)) ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Daminozide, Succinic acid 2,2-Dimethylhydrazide

D4015-25G D4015-5G
TCIAD4015-25GEA 171 EUR
TCIAD4015-25G TCIAD4015-5G
Daminozide (succinic acid mono(2,2-dimethylhydrazide)) ≥98.0% (by GC, titration analysis)
Daminozide (succinic acid mono(2,2-dimethylhydrazide))
Formula: C₆H₁₂N₂O₃
Punto di fusione: 155°C
MDL Number: MFCD00002787
CAS Number: 1596-84-5
EINECS: 216-485-9
Merck Index: 14,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4e693d5f6a98a8772e8e2b6b72920d2fc29da3&vid=ba4e693d5f6a98a8772e8e2b6b72920d2fc29da3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR