Stampa…

1,3-Diacetoxyacetone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,3-Diacetoxy-2-propanone, 2-Oxopropane-1,3-diyl diacetate

D2843-25G D2843-5G
TCIAD2843-25GEA 246 EUR
TCIAD2843-25G TCIAD2843-5G
1,3-Diacetoxyacetone ≥98.0% (by GC)
1,3-Diacetoxyacetone
Formula: C₇H₁₀O₅
Peso molecolare: 174,1513 g/mol
MDL Number: MFCD01310655
CAS Number: 6946-10-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR