Stampa…

(Methylthio)acetonitrile ≥99.0% (by GC)

Fornitore: TCI
Pericolo

M0941-10G M0941-25G
TCIAM0941-10GEA 65 EUR
TCIAM0941-10G TCIAM0941-25G
(Methylthio)acetonitrile ≥99.0% (by GC)
(Methylthio)acetonitrile
Formula: C₃H₅NS
Punto di ebollizione: 60°C
Flash Pt: 68°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00001925
CAS Number: 35120-10-6
EINECS: 252-383-0
UN: 1760
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4b01638dc2c068206ea9aca607783d1b2175a3d&vid=f4b01638dc2c068206ea9aca607783d1b2175a3d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR