N,N-Diphenylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₃H₁₀ClNO |
MDL Number:
MFCD00000633 CAS Number: 83-01-2 EINECS: 201-450-2 UN: 3261 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White to Dark blue powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...