Ethyl-1,3-dithiane-2-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D1648-1G
D1648-5G
TCIAD1648-1GEA
52
EUR
TCIAD1648-1G
TCIAD1648-5G
Ethyl-1,3-dithiane-2-carboxylate ≥98.0% (by GC)
Ethyl-1,3-dithiane-2-carboxylate
Formula:
C₇H₁₂O₂S₂ Peso molecolare: 192,30 g/mol Flash Pt: 55°C |
MDL Number:
MFCD00006657 CAS Number: 20462-00-4 EINECS: 243-838-4 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...