1-(Trifluoroacetyl)imidazole ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₅H₃F₃N₂O Punto di ebollizione: 83°C Flash Pt: 45°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00014501 CAS Number: 1546-79-8 EINECS: 216-282-5 UN: 1993 ADR: 3,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light red to Green clear liquid |
Purity (Nonaqueous Titration) | min. 98.0 % |
Effect test of GC derivalization | Effective as derivatization agent Dodecylamine |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...