Stampa…

1-(p-Toluenesulphonyl)imidazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole

T1985-25G T1985-5G
TCIAT1985-25GEA 135 EUR
TCIAT1985-25G TCIAT1985-5G
1-(p-Toluenesulphonyl)imidazole ≥98.0% (by HPLC, total nitrogen)
1-(p-Toluenesulphonyl)imidazole
Formula: C₁₀H₁₀N₂O₂S
Peso molecolare: 222,26 g/mol
MDL Number: MFCD00005285
CAS Number: 2232-08-8
EINECS: 218-771-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19d69d9e3ddc778219e4bb8b2d44a63833417b41&vid=19d69d9e3ddc778219e4bb8b2d44a63833417b41&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR