Stampa…

4-Methylumbelliferone ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-MU

M0453-25G M0453-500G M0453-100G
TCIAM0453-25GEA 43 EUR
TCIAM0453-25G TCIAM0453-500G TCIAM0453-100G
4-Methylumbelliferone ≥98.0% (by titrimetric analysis)
4-Metilumbelliferone
Formula: C₁₀H₈O₃
Peso molecolare: 176,17 g/mol
MDL Number: MFCD00006866
CAS Number: 90-33-5
EINECS: 201-986-7
Merck Index: 14,04854
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14bfb37c9f6212545504e79cf9d938b792e19c9&vid=b14bfb37c9f6212545504e79cf9d938b792e19c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR