Stampa…

7-(Diethylamino)-4-methylcoumarin ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Coumarin 1, 7-(Diethylamino)-4-methyl-2H-chromen-2-one, 7-Diethylamino-4-methylcoumarin

M0631-25G M0631-500G
TCIAM0631-25GEA 29 EUR
TCIAM0631-25G TCIAM0631-500G
7-(Diethylamino)-4-methylcoumarin ≥98.0% (by GC, titration analysis)
7-(Diethylamino)-4-methylcoumarin
Formula: C₁₄H₁₇NO₂
Punto di ebollizione: 243°C
MDL Number: MFCD00006864
CAS Number: 91-44-1
EINECS: 202-068-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47b8ea54b92023d92718633f89fd3532c71cea4f&vid=47b8ea54b92023d92718633f89fd3532c71cea4f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 74.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR