N,N-Dimethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
D0695-25G
D0695-500G
TCIAD0695-25GEA
17
EUR
TCIAD0695-25G
TCIAD0695-500G
N,N-Dimethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
N,N-Dimethylcarbamoyl chloride
Formula:
C₃H₆ClNO Flash Pt: 82°C |
MDL Number:
MFCD00000635 CAS Number: 79-44-7 EINECS: 201-208-6 UN: 2262 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...