Stampa…

1,3-Dichloro-2-iodobenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D3593-25G D3593-5G
TCIAD3593-25GEA 88 EUR
TCIAD3593-25G TCIAD3593-5G
1,3-Dichloro-2-iodobenzene ≥98.0% (by GC)
1,3-Dichloro-2-iodobenzene
Formula: C₆H₃Cl₂I
Peso molecolare: 272,90 g/mol
MDL Number: MFCD00060658
CAS Number: 19230-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab9071d132e0f9c9c13627d33a51330f11943322&vid=ab9071d132e0f9c9c13627d33a51330f11943322&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR