Stampa…

2-Bromoanthracene ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B2616-1G B2616-5G
TCIAB2616-1GEA 166 EUR
TCIAB2616-1G TCIAB2616-5G
2-Bromoanthracene ≥97.0% (by GC)
2-Bromoanthracene
Formula: C₁₄H₉Br
Peso molecolare: 257,1300 g/mol
MDL Number: MFCD07784002
CAS Number: 7321-27-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 218.0 to 222.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR