Stampa…

1-Bromoanthracene ≥97.0% (by GC)

Fornitore: TCI EUROPE

B2615-500MG B2615-100MG
TCIAB2615-500MGEA 215 EUR
TCIAB2615-500MG TCIAB2615-100MG
1-Bromoanthracene ≥97.0% (by GC)
1-Bromoanthracene
Formula: C₁₄H₉Br
MDL Number: MFCD08276282
CAS Number: 7397-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11688a605d21fd5a2662e89f85705a17e920d065&vid=11688a605d21fd5a2662e89f85705a17e920d065&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 97.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR