4,4,4-Trifluorobutyric acid ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
T2670-1G
T2670-5G
TCIAT2670-1GEA
22
EUR
TCIAT2670-1G
TCIAT2670-5G
4,4,4-Trifluorobutyric acid ≥98.0% (by GC)
4,4,4-Trifluorobutyric acid
Formula:
C₄H₅F₃O₂ Punto di ebollizione: 167°C Punto di fusione: 31°C |
MDL Number:
MFCD00077604 CAS Number: 406-93-9 UN: 3261 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...