Tetraethyl orthocarbonate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
T1107-25ML
T1107-5ML
TCIAT1107-25MLEA
104
EUR
TCIAT1107-25ML
TCIAT1107-5ML
Tetraethyl orthocarbonate ≥97.0% (by GC)
Tetraethyl orthocarbonate
Formula:
C₉H₂₀O₄ Peso molecolare: 192,26 g/mol Punto di ebollizione: 160°C Flash Pt: 52°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009221 CAS Number: 78-09-1 EINECS: 201-082-2 UN: 3271 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...