Stampa…

Sebacic acid dihydrazide ≥96.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: Decanedihydrazide

S0224-25G S0224-500G
TCIAS0224-25GEA 24 EUR
TCIAS0224-25G TCIAS0224-500G
Sebacic acid dihydrazide ≥96.0% (by titrimetric analysis)
Sebacic acid dihydrazide
Formula: C₁₀H₂₂N₄O₂
MDL Number: MFCD00039763
CAS Number: 925-83-7
EINECS: 213-126-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37b45fb2119871b8ba62bc967f09b10de0c314a9&vid=37b45fb2119871b8ba62bc967f09b10de0c314a9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 96.0 %
Melting point 185.0 to 191.0 deg-C
Solubility in hot DMSO very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR