2-Methoxyethyl acetoacetate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methoxyethyl 3-oxobutanoate
A1583-25G
A1583-500G
TCIAA1583-25GEA
17
EUR
TCIAA1583-25G
TCIAA1583-500G
2-Methoxyethyl acetoacetate ≥95.0% (by GC)
2-Methoxyethyl acetoacetate
Formula:
C₇H₁₂O₄ Peso molecolare: 160,1678 g/mol Punto di ebollizione: 120°C |
MDL Number:
MFCD00009648 CAS Number: 22502-03-0 EINECS: 245-043-8 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...