Stampa…

4,4'-Dinonyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4′-Dinonyl-2,2′-dipyridyl, 4,4'-Dinonyl-2,2'-bipyridine, 4,4'-Di-n-nonyl-2,2'-bipyridine

D3917-1G D3917-5G
TCIAD3917-1GEA 34 EUR
TCIAD3917-1G TCIAD3917-5G
4,4'-Dinonyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
4,4'-Dinonyl-2,2'-bipyridyl
Formula: C₂₈H₄₄N₂
Peso molecolare: 408,67 g/mol
MDL Number: MFCD00800915
CAS Number: 142646-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67e6307332cdbb21a542870eec7e27155758ad6d&vid=67e6307332cdbb21a542870eec7e27155758ad6d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR