Stampa…

3-(2-Furyl)acrylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Furan-2-yl)acrylic acid

F0084-10G F0084-25G
TCIAF0084-10GEA 66 EUR
TCIAF0084-10G TCIAF0084-25G
3-(2-Furyl)acrylic acid ≥98.0% (by GC, titration analysis)
3-(2-Furyl)acrylic acid
Formula: C₇H₆O₃
Peso molecolare: 138,12 g/mol
MDL Number: MFCD00003257
CAS Number: 539-47-9
EINECS: 208-718-8
Merck Index: 14,04297
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=189f1e453113f422df9cce7e0946c1b12da307cc&vid=189f1e453113f422df9cce7e0946c1b12da307cc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 143.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR