Stampa…

Tobramycin ≥94.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol

T2503-5G T2503-25G
TCIAT2503-5GEA 109 EUR
TCIAT2503-5G TCIAT2503-25G
Tobramycin ≥94.0% (by titrimetric analysis)
Tobramycin
Formula: C₁₈H₃₇N₅O₉
Peso molecolare: 467,51 g/mol
Punto di fusione: 178°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00077885
CAS Number: 32986-56-4
EINECS: 251-322-5
Merck Index: 14,09490
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c16df67a3dbd97fde6585a1d5058878fbda378a5&vid=c16df67a3dbd97fde6585a1d5058878fbda378a5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 94.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +138.0 to +148.0 deg (C=4, H2O) (calcd.on anh.substance)
Water max. 8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR